APC 2 FLASH Multirisk Labcoats
: ALSICO
Multirisk labcoat especially designed for APC 2.
- ATPV value: 26,6 cal/cm²
- ELIM value: 21 cal/cm²
- Modern fit
- Upright, rounded collar
- Centre front closure with hidden press studs, ECO design
- Inside zipped pocket
- 2 large side pockets with flap and hidden press studs
- Long sleeves with sleeve cuffs and elastic
- Piping in accent colour
Industrial launderable.
: Oeko-Tex® standard 100; EN 1149-5 2018; IEC 61482 - 2: 2018 - APC2; EN ISO 11612:2015 (A1, B1, C1, D0, E0, F1); EN 11611:2015 - Class 2 A1; EN 13034 +A1.
- Catalog No:
- ALSW1030929-4XL
- ALSW1030929-L
- ALSW1030929-XL
- ALSW1030929-2XL
- ALSW1030929-3XL
- ALSW1030929-M
- ALSW1030929-S
- Aplikace oblečení:
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Risk area:
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Rod:
- Pro muže
- Pro muže
- Pro muže
- Pro muže
- Pro muže
- Pro muže
- Pro muže
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Typ uzávěru:
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Tkanina:
- Outer fabric: 54% modacrylic/44% cotton/2% antistatic
Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon - Outer fabric: 54% modacrylic/44% cotton/2% antistatic
Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon - Outer fabric: 54% modacrylic/44% cotton/2% antistatic
Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon - Outer fabric: 54% modacrylic/44% cotton/2% antistatic
Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon - Outer fabric: 54% modacrylic/44% cotton/2% antistatic
Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon - Outer fabric: 54% modacrylic/44% cotton/2% antistatic
Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon - Outer fabric: 54% modacrylic/44% cotton/2% antistatic
Lining: 45% modacrylic/35% cotton/18% polyamide/2% carbon
- Hmotnost:
- 480 g/m²
- 480 g/m²
- 480 g/m²
- 480 g/m²
- 480 g/m²
- 480 g/m²
- 480 g/m²
Frequently Bought Together