Concept Collection™ Open Top Waste/Recycling Bin
Katalogové číslo 129-1951
Dodavatel: Rubbermaid Commercial Products
Specifikace
- Ø×H:291×356 mm
- Kapacita:19 L
- Barva:Stříbrná
- Materiál:Kov
- Bal.:1
Specifikace
O tomto produktu
Space-efficient, economical, easy and effective way to recycle.
- No snag curled rim protects clothing
- Baked-on epoxy finish
- Solid steel base
- Vinyl base ring protects floors
- Includes a five year warranty against normal wear and tear
A designer solution for deskside disposal.