Orders LinkContactUsLinkComponent
KoolPlate™ Cooling Tray
KoolPlate™ Cooling Tray
KoolPlate™ 6 cooling trays with 1 insulator base, 279,4×139,7×25,4 mm
Katalogové číslo SIMPM479-24
Dodavatel:   Simport Scientific
KoolPlate™ Cooling Tray
Katalogové číslo SIMPM479-24
Dodavatel:  Simport Scientific
KoolPlate™ Cooling Tray
KoolPlate™ Cooling Tray
KoolPlate™ 6 cooling trays with 1 insulator base, 279,4×139,7×25,4 mm
Přejít na:
Price
8 200,00 Kč
Přihlásit se pro dostupnost
1 * 6 KS
Product Details & Documents
The Simport KoolPlate™ is ideal for chilling and preparing paraffin blocks, for sectioning at the microtome or for performing other laboratory procedures.
  • For chilling and preparing paraffin blocks
  • Keeps paraffin blocks cold up to 4 hours
  • Tray made of high impact polystyrene for longevity
  • Insulator base made of styrofoam
The KoolPlate™ freezes overnight (16 hours) in a conventional freezer and the surface remains cold for up to 4 hours when used with the supplied styrofoam insulator base placed under the KoolPlate™. It will prolong cooling and prevent moisture accumulation. It is made of thick wall high impact polystyrene in order to ensure a sturdy construction that will last for years.
The KoolPlate™ is stackable, a great space-saving feature. Warning–Not for use with organic solvents such as toluene, xylene, acetone, etc.
Caution: Not for use with organic solvents such as toluene, xylene, acetone, etc.
Specifikace
  • Popis:
    KoolPlate™ 6 cooling trays with 1 insulator base
  • Š×H×V:
    279,4×139,7×25,4 mm