Specifikace
- Popis:AdFlo™ Powered air respirator filter and battery kit
- Bal.:1 KIT
Specifikace
O tomto produktu
AdFlo™ Powered air respirator filter and battery kit contains the Adflo™ filter and Adflo™ heavy-duty Li-ion battery. It’s designed for use with 3M™ Adflo™ powered air respirator.
- Replacement powered air respirator and battery
- Upgrade kit for Adflo™ Powered air respirator
- Captures organic, inorganic, and acid gases and vapours
- Always to be used together with particle filter
- Heavy-duty battery fully charges in 5,5 hours and has an operating time of 10 to 12 hours
Get reliable protection from gases and vapours by upgrading the 3M™ Adflo™ Powered Air Respirator with an AdFlo™ Powered Air Respirator Filter and Battery Kit. The kit includes the high-capacity Li-ion battery that extends the operating hours of the respirator.
The gas filter must always be used together with the particle filter (837010). Gas filters should be replaced according to workplace requirements. It is not recommended to wait until odours can be detected. In some configurations it is also required to use the breathing tube with silencer (834009). Always verify approved combinations with a Safety Engineer or contact a certified Industrial Hygienist for advice.