Accessories for HAEMAFlex ™ Swinging Bucket Rotor 6/8
Catalog # HERA75003857
Supplier: Thermo Scientific
Specifications
- Kapacita:6/8×500 ml
- Popis:Triple or Quad blood bag adapter, 450 - 500 ml
- Bal.:1
- Určeno pro:Single blood bag bucket with filter pack
Specifications
About this item
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.