Cole-Parmer® SH-200 See-Saw Rocking Shakers
Gently mix live cultures.
- Non-slip mat and platform lip hold vessels securely in place
- Built-in timer and digital speed LED display provide reproducible results
- Can be used in incubators
These see-saw rocking shakers provide a gentle rocking action at a 7° tilt angle, perfect for culture flasks and Petri dishes. Easily set and view the speed and the timer on the LED display. Control speed from 5 to 70 min⁻¹. The microprocessor maintains speed accuracy even over long runs. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert.
Durable shakers are rated for continuous use. Use in incubators and environmental or cold chambers up to 40 °C.
: Units include flat platform, non-slip mat, and power cord.
- Catalog No:
- STUA51901-14
- STUA51901-06
- Model:
- Mini
- Large
- Rozsah rychlosti:
- 5 - 70 min⁻¹
- 5 - 70 min⁻¹
- Úhel náklonu:
- 7°
- 7°
- Š×H×V:
- 275×335×155 mm
- 390×450×165 mm
- Plošina ŠxH:
- 235×235 mm
- 355×355 mm
- Displej:
- LED
- LED
- Maximální náplň:
- 3 kg
- 10 kg
- Časovač:
- 1 - 999 min
- 1 - 999 min
Frequently Bought Together