b.safe Blind Fitting PFAS Analytic
Catalog # BOHLM401-01
Supplier: Bohlender
Specifications
- Závit:UNF ¹/₄"
- Popis:Blind fitting PFAS analytic, 10 per pack
- Bal.:1
Specifications
About this item
To close unused connections on b.safe Caps PFAS Analytic.
- Made of PE
- For PFAS analysis
- With ergonomically shaped grip surface
- easy mounting
Blind Fittings made of PE for tight closure of unused connectors on b.safe Caps PFAS Analytic. With ergonomic gripping surface for easy assembly.