Order Entry
Czech Republic
ContactUsLinkComponent
Anti-C4 binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-C4 binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-15184R-A350
Supplier:  Bioss
Anti-C4 binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-15184R-A350
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    C4 binding protein
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 350
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Epitop:
    1-100/252
  • Formulář:
    Liquid
  • Synonyma antigenu:
    PRP|OTTHUMP00000034375|C4b-binding protein beta chain|OTTHUMP00000034288|OTTHUMP00000034289|OTTHUMP0000003474|Complement component 4-binding protein beta|Complement component 4 binding protein beta chain|C4BP|Complement component 4 binding protein beta|C4b binding protein beta chain|C4BPB_HUMAN|Proline rich protein|C4BPB
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molekulární hmotnost:
    26 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human C4 binding protein
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

C4BPB is a member of a superfamily of proteins composed predominantly of tandemly arrayed short consensus repeats of approximately 60 amino acids. A single, unique beta-chain encoded by this gene assembles with seven identical alpha-chains into the predominant isoform of C4b-binding protein, a multimeric protein that controls activation of the complement cascade through the classical pathway. C4b-binding protein has a regulatory role in the coagulation system also, mediated through the beta-chain binding of protein S, a vitamin K-dependent protein that serves as a cofactor of activated protein C. The genes encoding both alpha and beta chains are located adjacent to each other on human chromosome 1 in the regulator of complement activation gene cluster. Alternative splicing gives rise to multiple transcript variants

Type: Primary
Antigen: C4 binding protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 1-100/252
Host: Rabbit
Isotype: IgG
Reactivity: Human