Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MPP7 Rabbit Polyclonal Antibody (Cy3®)
Anti-MPP7 Rabbit Polyclonal Antibody (Cy3®)
Katalogové číslo BOSSBS-13737R-CY3
Dodavatel:  Bioss
Anti-MPP7 Rabbit Polyclonal Antibody (Cy3®)
Katalogové číslo BOSSBS-13737R-CY3
Dodavatel:  Bioss

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    MPP7
  • Klonovalita:
    Polyclonal
  • Spojení:
    Cy3®
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Epitop:
    301-400/576
  • Formulář:
    Liquid
  • Synonyma antigenu:
    FLJ32798|MAGUK p55 subfamily member 7|Palmitoylated membrane protein 7|Membrane protein palmitoylated 7 MAGUK p55 subfamily member 7
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molekulární hmotnost:
    65 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human MPP7
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifikace

O tomto produktu

The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression, and signaling. The MAGUK family is divided into four subfamilies: DLG-like, ZO1-like, p55-like and LIN2-like. MPP7 (MAGUK p55 subfamily member 7), also known as palmitoylated membrane protein 7, is a 576 amino acid protein that belongs to the MAGUK family and assists in the assembly of protein complexes. A peripheral membrane protein that exists as a heterodimer, MPP7 enhances tight junction formation and epithelial cell polarity. MPP7 contains one SH3 domain, a guanylate kinase-like domain, two L27 domains and a single PDZ (DHR) domain. The gene encoding MPP7 maps to human chromosome 10p11.23

Type: Primary
Antigen: MPP7
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 301-400/576
Host: Rabbit
Isotype: IgG
Reactivity: