Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IKK alpha/beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-7557R-A680
 :  Bioss
Anti-IKK alpha/beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-7557R-A680
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    IKK alpha / beta
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 680
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    101-200/756
  • Formulář:
    Liquid
  • Synonyma antigenu:
    NFKBIKA|I Kappa B kinase alpha|IKK alpha+IKK beta|IKK beta|IKKA_HUMAN|IkBKB|I Kappa B kinase beta|IKK alpha|IKKB|I kappa B kinase 1|IKK1|IKK2|IKKA|NFKBIKB|I kappa B kinase 2|IkBKA|IKKB_HUMAN
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    83 kDa
  • Skladovací teplota:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human IKK alpha + IKK beta
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

 

 

IKK is a serine protein kinase, and the IKK complex contains alpha and beta subunits (IKK alpha and IKK beta). IKK alpha and IKK beta interact with each other and both are essential for NFkB activation.

Type: Primary
Antigen: IKK alpha / beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/756
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat