Specifikace
- Druh protilátky:Primární
- Název antigenu:FGFR5
- Klonovalita:Polyclonal
- Spojení:Alexa Fluor® 750
- Hostitel:Rabbit
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Izotyp:IgG
- Western blot:Yes
- Epitop:231-330/504
- Formulář:Liquid
- Identifikační číslo genu:53834
- Synonyma antigenu:FGFR like protein|FGFR5|Fibroblast growth factor receptor 5|FGF receptor like protein 1|FGF homologous factor receptor|FGFR 5|FGRL1_HUMAN|Fibroblast growth factor receptor like 1|FHFR|FGFRL1
- Modifikace:Unmodified
- Skladovací pufr:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molekulární hmotnost:52 kDa
- Skladovací teplota:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Koncentrace:1 µg/µl
- Transportní teplota:4 °C
- Imunogen:KLH conjugated synthetic peptide derived from human FGFR5/FGFRL1
- Testované aplikace:ICC
- Čištění:Purified by Protein A
- Typ balení:Vial
- Bal.:100 µl
Specifikace
O tomto produktu
FGFRL1 is a member of the fibroblast growth factor receptor (FGFR) family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. A marked difference between FGFRL1 and the other family members is its lack of a cytoplasmic tyrosine kinase domain. The result is a transmembrane receptor that could interact with other family members and potentially inhibit signaling. Multiple alternatively spliced transcript variants encoding the same isoform have been found.
Type: Primary
Antigen: FGFR5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 231-330/504
Host: Rabbit
Isotype: IgG
Reactivity: