Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-INTS3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9766R-A680
 :  Bioss
Anti-INTS3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9766R-A680
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    INTS3
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 680
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    901-1043/1043
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    65123
  • Synonyma antigenu:
    INTS3|INT3|C1orf193|Integrator complex subunit 3|SOSSA|Sensor of ssDNA subunit A|SOSS complex subunit A|Sensor of single-strand DNA complex subunit A|C1orf60|SOSS-A
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    118 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human INTS3
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

 

 

Component of the Integrator complex. The Integrator complex is involved in the small nuclear RNAs (snRNA) U1 and U2 transcription and in their 3'-box-dependent processing. The Integrator complex is associated with the C-terminal domain (CTD) of RNA polymerase II largest subunit (POLR2A) and is recruited to the U1 and U2 snRNAs genes. Component of the SOSS complex, a multiprotein complex that functions downstream of the MRN complex to promote DNA repair and G2/M checkpoint. The SOSS complex associates with single-stranded DNA at DNA lesions and influences diverse endpoints in the cellular DNA damage response including cell-cycle checkpoint activation, recombinational repair and maintenance of genomic stability. The SOSS complex is required for efficient homologous recombination-dependent repair of double-strand breaks (DSBs) and ATM-dependent Signalling pathways. In the SOSS complex, it is required for the assembly of the complex and for stabilisation of the complex at DNA damage sites.

Type: Primary
Antigen: INTS3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 901-1043/1043
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat