Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-RGS4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9452R-A680
 :  Bioss
Anti-RGS4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-9452R-A680
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    RGS4
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 680
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    51-150/205
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    5999
  • Synonyma antigenu:
    Schizophrenia disorder 9|MGC60244|RGP 4|MGC2124|Regulator of G-protein signaling 4|RGS4_HUMAN|SCZD9|RGS4|RGS 4|Regulator of G protein signalling 4|RGP-4|SCZD 9
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human RGS4
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

 

 

Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Activity on G(z)-alpha is inhibited by phosphorylation of the G-protein. Activity on G(z)-alpha and G(i)-alpha-1 is inhibited by palmitoylation of the G-protein.

Type: Primary
Antigen: RGS4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/205
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat