Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MEK1 Thr286 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-MEK1 Thr286 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3269R-A750
Supplier:  Bioss
Anti-MEK1 Thr286 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3269R-A750
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    MEK1 Thr286
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 750
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    5604
  • Synonyma antigenu:
    MKK 1|Dual specicity mitogen activated protein kinase kinase 1|Mitogen activated protein kinase kinase 1|MAPKK 1|Protein kinase mitogen activated kinase 1|Protein kinase mitogen activated, kinase 1|MAPKK1|MP2K1_HUMAN|MAP kinase/Erk kinase 1|MEK 1|Protein kinase mitogen activated kinase 1 MAP kinase kinase 1|PRKMK 1|ERK activator kinase 1|PRKMK1|Map2K1|MAP kinase kinase 1|MAPK/ERK kinase 1|MEKK1|MKK1|MAP/Erk kinase 1
  • Modifikace:
    Phosphorylation (Thr286)
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    43 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic phosphopeptide derived from human MEK1 around the phosphorylation site of Thr286
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

The protein encoded by this gene is a member of the dual specificity protein kinase family, which acts as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein kinase lies upstream of MAP kinases and stimulates the enzymatic activity of MAP kinases upon wide variety of extra- and intracellular signals. As an essential component of MAP kinase signal transduction pathway, this kinase is involved in many cellular processes such as proliferation, differentiation, transcription regulation and development.

Type: Primary
Antigen: MEK1 Thr286
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat