ContactUsLinkComponent
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-2197R-A750
 :  Bioss
Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-2197R-A750
 :  Bioss
  • Druh protilátky:
    Primární
  • Název antigenu:
    duck hepatitis A virus 1 Genome polyprotein
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 750
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Virus
  • Western blot:
    Yes
  • Epitop:
    851-950/2249
  • Formulář:
    Liquid
  • Synonyma antigenu:
    DHAV|polyprotein DHAV|polyprotein [duck hepatitis A virus]
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    252 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    MAP peptide derived from polyprotein [duck hepatitis A virus] FC64/CHN/2009
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl
Duck hepatitis A virus genome polyprotein.
Type: Primary
Antigen: duck hepatitis A virus 1 Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 851-950/2249
Host: Rabbit
Isotype: IgG
Reactivity: Virus, Others