Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MEK1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-MEK1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1041R-A680
Supplier:  Bioss
Anti-MEK1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1041R-A680
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    MEK1/2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 680
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    301-393/393
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    5604
  • Synonyma antigenu:
    MEK1|MAPKK 2|MEK 2|MAP kinase kinase 2|Dual specicity mitogen-activated protein kinase kinase 1|MAP2K1|MAPKK1|ERK activator kinase 2|ERK activator kinase 1|PRKMK1|MAP kinase kinase 1|MAPK/ERK kinase 1|MKK1
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    43 kDa
  • Skladovací teplota:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human MEK1
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

MAP2K1
Dual specificity protein kinase which acts as an essential component of the MAP kinase signal transduction pathway. Binding of extracellular ligands such as growth factors, cytokines and hormones to their cell-surface receptors activates RAS and this initiates RAF1 activation. RAF1 then further activates the dual-specificity protein kinases MAP2K1/MEK1 and MAP2K2/MEK2. Both MAP2K1/MEK1 and MAP2K2/MEK2 function specifically in the MAPK/ERK cascade, and catalyse the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in the extracellular signal-regulated kinases MAPK3/ERK1 and MAPK1/ERK2, leading to their activation and further transduction of the signal within the MAPK/ERK cascade. Depending on the cellular context, this pathway mediates diverse biological functions such as cell growth, adhesion, survival and differentiation, predominantly through the regulation of transcription, metabolism and cytoskeletal rearrangements. One target of the MAPK/ERK cascade is peroxisome proliferator-activated receptor gamma (PPARG), a nuclear receptor that promotes differentiation and apoptosis. MAP2K1/MEK1 has been shown to export PPARG from the nucleus. The MAPK/ERK cascade is also involved in the regulation of endosomal dynamics, including lysosome processing and endosome cycling through the perinuclear recycling compartment (PNRC), as well as in the fragmentation of the Golgi apparatus during mitosis.

MAP2K2
catalyses the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in MAP kinases. Activates the ERK1 and ERK2 MAP kinases.

Type: Primary
Antigen: MEK1/2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-393/393
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others