Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0995R-A750
Supplier:  Bioss
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0995R-A750
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Adrenomedullin
  • Symbol antigenu:
    ADM
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 750
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Rat
  • Epitop:
    1-100/185
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    133
  • Synonyma antigenu:
    ADML_HUMAN.|AM|Proadrenomedullin N-20 terminal peptide|Adrenomedullin|ADM|PAMP|ProAM N-terminal 20 peptide|Contains|RecName|ProAM-N20
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    20 kDa
  • Skladovací teplota:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human ADM
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterised by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is up-regulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.

Type: Primary
Antigen: Adrenomedullin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/185
Host: Rabbit
Isotype: IgG
Reactivity: Rat