Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PSEN1 Mouse Polyclonal Antibody (Alexa Fluor® 750)
Anti-PSEN1 Mouse Polyclonal Antibody (Alexa Fluor® 750)
Katalogové číslo BOSSBS-0024M-A750
Supplier:  Bioss
Anti-PSEN1 Mouse Polyclonal Antibody (Alexa Fluor® 750)
Katalogové číslo BOSSBS-0024M-A750
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Presenilin-1
  • Symbol antigenu:
    PSEN1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 750
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Mouse
  • Epitop:
    10-80/467
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    29192
  • Synonyma antigenu:
    Psnl1|Psen1|Presenilin-1|Protein S182|PS-1
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    34/52 kDa
  • Skladovací teplota:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from rat presenilin 1
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

Probable catalytic subunit of the gamma-secretase complex, an endoprotease complex that catalyses the intramembrane cleavage of integral membrane proteins such as Notch receptors and APP (beta-amyloid precursor protein). Requires the other members of the gamma-secretase complex to have a protease activity. May play a role in intracellular signaling and gene expression or in linking chromatin to the nuclear membrane. Stimulates cell-cell adhesion though its association with the E-cadherin/catenin complex. Under conditions of apoptosis or calcium influx, cleaves E-cadherin promoting the disassembly of the E-cadherin/catenin complex and increasing the pool of cytoplasmic beta-catenin, thus negatively regulating Wnt signaling. May also play a role in hematopoiesis (By similarity).

Type: Primary
Antigen: Presenilin-1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 10-80/467
Host: Mouse
Isotype: IgG
Reactivity: Mouse