Order Entry
Czech Republic
ContactUsLinkComponent
Anti-c-Raf Ser339 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-c-Raf Ser339 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5652R-A680
Supplier:  Bioss
Anti-c-Raf Ser339 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5652R-A680
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    c-Raf Ser339
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 680
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    5894
  • Synonyma antigenu:
    Raf-1|RAF1|CRAF|CMD1NN|RAF|NS5|RAF proto-oncogene serine/threonine-protein kinase|c-Raf|Proto-oncogene c-RAF
  • Modifikace:
    Phosphorylation (Ser339)
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    73 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic phosphopeptide derived from human c-Raf around the phosphorylation site of Ser339
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

Serine/threonine-protein kinase that acts as a regulatory link between the membrane-associated Ras GTPases and the MAPK/ERK cascade, and this critical regulatory link functions as a switch determining cell fate decisions including proliferation, differentiation, apoptosis, survival and oncogenic transformation. RAF1 activation initiates a mitogen-activated protein kinase (MAPK) cascade that comprises a sequential phosphorylation of the dual-specific MAPK kinases (MAP2K1/MEK1 and MAP2K2/MEK2) and the extracellular signal-regulated kinases (MAPK3/ERK1 and MAPK1/ERK2). The phosphorylated form of RAF1 (on residues Ser-338 and Ser-339, by PAK1) phosphorylates BAD/Bcl2-antagonist of cell death at 'Ser-75'. Phosphorylates adenylyl cyclases: ADCY2, ADCY5 and ADCY6, resulting in their activation. Phosphorylates PPP1R12A resulting in inhibition of the phosphatase activity. Phosphorylates TNNT2/cardiac muscle troponin T. Can promote NF-kB activation and inhibit signal transducers involved in motility (ROCK2), apoptosis (MAP3K5/ASK1 and STK3/MST2), proliferation and angiogenesis (RB1). Can protect cells from apoptosis also by translocating to the mitochondria where it binds BCL2 and displaces BAD/Bcl2-antagonist of cell death. Regulates Rho signaling and migration, and is required for normal wound healing. Plays a role in the oncogenic transformation of epithelial cells via repression of the TJ protein, occludin (OCLN) by inducing the up-regulation of a transcriptional repressor SNAI2/SLUG, which induces down-regulation of OCLN. Restricts caspase activation in response to selected stimuli, notably Fas stimulation, pathogen-mediated macrophage apoptosis, and erythroid differentiation.

Type: Primary
Antigen: c-Raf Ser339
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse