Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ITM5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Anti-ITM5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Katalogové číslo BOSSBS-15518R-A647
Dodavatel:  Bioss
Anti-ITM5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Katalogové číslo BOSSBS-15518R-A647
Dodavatel:  Bioss

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    ITM5
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 647
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Western blot:
    Yes
  • Epitop:
    1-100/132
  • Formulář:
    Liquid
  • Synonyma antigenu:
    Interferon-induced transmembrane protein 5|Hrmp1|M5_HUMAN|BRIL|Bone-restricted interferon-induced transmembrane protein-like protein|Fragilis4|ITM5
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekulární hmotnost:
    14 kDa
  • Skladovací teplota:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human IFITM5
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifikace

O tomto produktu

IFITM5 is a membrane protein thought to play a role in bone mineralisation. This gene is located on chromosome 11 in a cluster of related genes which are induced by interferon, however, this gene has not been shown to be interferon inducible. A similar gene, located in a gene cluster on mouse chromosome 7, is a member of the interferon-inducible fragilis gene family. The mouse gene encodes a transmembrane protein described as participating in germ cell competence. A mutation in the 5' UTR of this gene has been associated with osteogenesis imperfecta type V (PMID: 22863190, 22863195).

Type: Primary
Antigen: ITM5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 1-100/132
Host: Rabbit
Isotype: IgG
Reactivity: