Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-CATSPER3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-CATSPER3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7136R-A750
Supplier:  Bioss
Anti-CATSPER3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7136R-A750
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    CATSPER3
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 750
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Western blot:
    Yes
  • Epitop:
    201-300/398
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    347732
  • Synonyma antigenu:
    Cation channel, sperm associated 3|CTSR3_HUMAN|CATSPER3|cav-like protein|Cav-like protein|CACRC|cation channel sperm-associated protein 3|calcium channel repeat containing 1|putative ion channel CatSper3|one-repeat calcium channel-like protein
  • Modifikace:
    Unmodified
  • Skladovací pufr:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molekulární hmotnost:
    46 kDa
  • Skladovací teplota:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentrace:
    1 µg/µl
  • Transportní teplota:
    4 °C
  • Imunogen:
    KLH conjugated synthetic peptide derived from human CATSPER3
  • Testované aplikace:
    ICC
  • Čištění:
    Purified by Protein A
  • Typ balení:
    Vial
  • Bal.:
    100 µl

Specifications

About this item

CatSpers (cation channel, sperm associated proteins) are ion transport proteins located on the surface of sperm cells in the principal piece of the sperm tail. CatSpers are vital to sperm motility, fertilization and cAMP-mediated calcium influx in sperm. There are four CatSper proteins in mammalian sperm, namely CatSper (or CatSper1), CatSper2, CatSper3 and CatSper4. CatSper proteins contain a single, six-transmembrane-spanning segment and exhibit the voltage-dependent Ca2+ channel four-repeat structure. CatSper proteins are believed to assemble into a heterotetrameric complex, forming an alkalinization-activated Ca2+-selective channel. Mutations in any of the genes encoding CatSper family proteins can result in male infertility. CatSper3 plays an important role in the hyperactivated motility of sperm cells, a process that is required in the preparation of sperm for fertilization.

Type: Primary
Antigen: CATSPER3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/398
Host: Rabbit
Isotype: IgG
Reactivity: