Anti-COL5A1-COL5A3 Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals
Certificates
About this item
Specifications
- Catalog No:
- ROCK600-401-107-05
- ROCK600-401-107-01
- Symbol antigenu:
- COL5A1-COL5A3
- COL5A1-COL5A3
- Název antigenu:
- Collagen alpha-1(V) chain
- Collagen alpha-1(V) chain
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Reaktivita:
- Human, Bovine
- Human, Bovine
- Hostitel:
- Rabbit
- Rabbit
- Identifikační číslo genu:
- 50509
- 50509
- Imunochemie:
- Yes
- Yes
- Imunogen:
- Collagen Type V from human and bovine placenta
- Collagen Type V from human and bovine placenta
- Formát:
- IgG
- IgG
- Formulář:
- Liquid (sterile filtered)
- Liquid (sterile filtered)
- Koncentrace:
- 1.0 mg/mL by UV absorbance at 280 nm
- 1.0 mg/mL by UV absorbance at 280 nm
- Skladovací pufr:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Skladovací teplota:
- 4°C
- 4°C
- Transportní teplota:
- 20
- 20
- Testované aplikace:
- Dot Blot
- Dot Blot
Specifications
Related Information
Frequently Bought Together