ContactUsLinkComponent
Arginase I (ARG1) Inhibitor Screening Kit (Colorimetric)
Arginase I (ARG1) Inhibitor Screening Kit (Colorimetric)
Catalog # ABCAAB283402-100
Supplier:  Abcam
CAS Number:  
undefined
Arginase I (ARG1) Inhibitor Screening Kit (Colorimetric)
Catalog # ABCAAB283402-100
Supplier:  Abcam
CAS Number:  
Specifications
  • Bal.:
    100 Tests
  • Popis:
    Arginase I inhibitor screening kit
Specifications
About this item
The most sensitive kit to evaluate potential human Arginase I inhibitors.
  • Simple and quick
  • Used to identify and characterize ARG1 inhibitors in a high-throughput format
Arginase (EC 3.5.3.1) is a manganese-containing enzyme which catalyzes the conversion of arginine into urea and ornithine, which is the final reaction in the urea cycle. Arginase I (ARG1) is the liver isoform of arginase. Recent studies showed that ARG1 expression by mature myeloid cells in tumor environment as demonstrated in a 3LL murine lung carcinoma model causes L-Arginine depletion by tumor-associated myeloid cells (TAMC).
L-arginine depletion suppresses immune-response against tumor cells due to inhibition to T-cell proliferation. In addition, the depletion of arginine increases the reactive nitrogen species (NOS) and reactive oxygen species (ROS), which, in consequence, induce T-cell apoptosis and supports antigenic cell proliferation.
Arginase I (ARG1) Inhibitor Screening Kit is designed for screening ARG1 inhibitors. Two substituted-2-amino-6-hexanoic acids have been studied as arginase inhibitor and in this kit, Amino-2-Borono-6-Hexanoic Acid (ABH) is provided as a positive control. The ARG1 activity is monitored by the increase in absorbance readings (OD 450 nm), while potential inhibitors will cause a decrease of absorbance.
Caution:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.