Cole-Parmer® Generator Probe Tool Kit for Homogenisers HO-200 and HO-400
Katalogové číslo STUA78905-47
Dodavatel: Cole Parmer
Specifikace
- Popis:Generator probe tool kit for HO-200 and HO-400 series homogenisers
- Bal.:1
Specifikace
O tomto produktu
Keep your homogeniser up and running.
- Optimise performance of your homogeniser generator probes
Ideal replacement parts for Cole-Parmer branded homogenisers and probes.
Informace o dodávce: Supplied with knife tool to remove 5, 7, and 10 mm probe knives, bearing removal tools for 5 and 7 mm generator probes and a shaft holder.