GCQ CleanType® Medical Silicone Keyboard with Illumination
Catalog # GETTKG24229
Supplier: GETT GERATETECHNIK
Specifications
- Popis:TKG-110-GCQ-MED, IP 68, USB, backlit, white, DE layout
- Š×H×V:380×130×13 mm
- Bal.:1
Specifications
About this item
The CleanType® Medical keyboard is specially manufactured for medical environments. With its versatility, it can be used in almost all areas of hospitals, clinics, and medical practices.
- IP 68 rated medical keyboard
- Fully washable and disinfectable
- Water-resistant silicone USB protection cap
- IEC 60601-1-2 certified
- For use in various areas: hospitals, offices, laboratories, production areas
The CleanType® Medical keyboard is completely waterproof, can be disinfected, and features integrated backlighting. The keyboard is a product of the GCQ series from GETT.
Certifications: CE, FCC, REACH.
Packaging: Keyboards are packed and delivered in environmentally friendly brown cardboard.