Cleanroom Boots, with Zipper
Supplier: ALSICO HIGHTECH
Certificates
About this item
This boot is made of Alsistrong® (98% polyester and 2% carbon). Suitable together with a coverall for the highest cleanroom classes ISO 4 to 9 or GMP classes A to D.
- Strap and studs at the top
- Studs to attach boot to the coverall
- Zipper in the front
- Coloured strap with 2-piece buckle over the foot (strap colour per size)
- Antistatic sole Combi P
- Reinforcement tape around the sole
Usable in ISO 4 to 9.
Certifications: IEST-RP-CC003.5.
Specifications
- Catalog No:
- ALSO1034488-S
- ALSO1034204-M
- ALSO1034204-SM
- ALSO1034204-L
- ALSO1034488-M
- ALSO1034488-SM
- ALSO1034204-ML
- ALSO1034204-XXL
- ALSO1034488-L
- ALSO1034488-ML
- ALSO1034204-XL
- ALSO1034488-XXL
- ALSO1034204-3XL
- ALSO1034488-XL
- ALSO1034204-XS
- ALSO1034488-3XL
- ALSO1034488-XS
- ALSO1034204-S
- Aplikace oblečení:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Rod:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Tkanina:
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Alsistrong® (98% polyester, 2% carbon)
- Hmotnost:
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²
- 118 g/m²