VWR®, Polypropylene Microplates, 96-Well
Katalogové číslo 548-3375
Dodavatel: VWR Collection
Specifikace
- Barva:Přírodní
- Popis:Polypropylene deep well microplate, Ø×H: 7×44 mm, well depth: 44 mm, square shaped
- Environmentally Preferable:
- Bal.:5
- Well volume:2000 µl
Specifikace
O tomto produktu
VWR® 96-Well plates and covers are designed for chromatography and high throughput screening (HTS) applications.
- SBS and ANSI standard footprint
- Low binding virgin PP for homogenous assays and storage
- –80 to +121 °C tolerant
VWR® 96-Well Microplates are manufactured specifically for chromatography. Multiple volumes and interior finishes allow for enhanced sample recovery. Low binding virgin polypropylene plates are perfect for high throughput screening (HTS), and sensitive samples.