Dual-wire wrist strap parking station, MagSnap 360™
Catalog # 115-3046
Supplier: DESCO
Specifications
- Popis:MagSnap 360™ Dual-wire wrist strap parking station
- Bal.:1
Specifications
About this item
This provides a storage terminal for the wrist cord when the workstation is unoccupied.
- Mounts to any surface conveniently located nearby the operator
- Spares operator jacks from the mechanical wear and tear of constant insertion and removal of the wrist cord
- Monitors display a PASS condition for the operator when the wrist cord is parked
Gross weight: 0,01 kg.