Surface and mat cleaner, Reztore®
Katalogové číslo 115-3048
Dodavatel: DESCO
Specifikace
- Popis:Surface and mat cleaner, 950 ml
- Balené:Reusable spray bottle
- Bal.:1
Specifikace
O tomto produktu
Reztore® Surface and mat cleaners removes dust, grease, grime, fingerprints, solder flux and other contaminants from ESD mats and other surfaces.
- One quart trigger spray bottle
- New alcohol-free formula
- Non-hazardous, non-flammable green chemistry formula
- No silicone or insulative residues
No special handling requirements for shipping and storage; may be used in normal workplace environments. Cleaners with silicone leave an insulative layer preventing conductive or dissipative mats from functioning properly.
Certifications: Lead-free RoHS compliant.