VWR®, Multilayer Cell Culture Flasks, 3 Layer, 5 Layer
Supplier: VWR Collection
Certificates
About this item
Multilayer cell culture flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive.
- Even distribution of media across all layers for homogenous cell growth
- Flexible design allows to pour or aspirate/recover cells using a pipet
- Consistent surface treatment for predictable scale-up
- Molded graduations stand out distinctly on these optically clear
- DNase/RNase free and non-pyrogenic
- Sterilized by irradiation SAL 10⁻⁶ (ISO 11137)
- Shelf Life: 3 years after month of production
Vent caps incorporate a 0,22 µm hydrophobic membrane for optimal gas exchange while protecting against contamination.
*Recommended working volume per layer.
Certifications: Manufactured in a Class 100,000 room environment under ISO 13485 and ISO 9001 quality management system.
Specifications
- Catalog No:
- 734-3418
- 734-3416
- 734-3417
- 734-3415
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M