Order Entry
Czech Republic
ContactUsLinkComponent
Trimesic acid trimethyl ester ≥98%
Trimesic acid trimethyl ester ≥98%
Catalog # ABCRAB133707.0025
Supplier:  abcr
CAS Number:  
Trimesic acid trimethyl ester ≥98%
Catalog # ABCRAB133707.0025
Supplier:  abcr
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle for solids
  • Bal.:
    25 g
  • Sekvence:
    3308

Specifications

About this item

 :
Boiling Pt:225…230 °C (0,5 mmHg)
Melting Pt:144…147 °C
Storage Temperature:Ambient
EINECS:220-215-5
CAS Number:2672-58-4
Formula:C₁₂H₁₂O₆
MDL Number:MFCD00008434