


Toluen
Catalog # MACH20056AT0M0900
Supplier: MACH
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1.000 mL
- Sekvence:2865
Specifications
About this item
:
Merck Index:13,09607
Boiling Pt:110,6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0,867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512