Temperature and humidity logger, EBI 20-TH1
: EBRO
Standard temperature and humidity data logger with internal humidity sensor and a memory capacity of 40000 measurements.
- Automatic recording of temperature data
- No network connection required
- Programming and evaluation with PC
- LED alarm
- Min/max values on display
- Intervall: 1 min to 24 h
EBI 20 data loggers are suitable for the continuous documentation and monitoring of temperature and humidity.
Applications: Transport monitoring, storage monitoring and process monitoring.
: Factory calibration certificate (with values).
: All EBI 20 data loggers are delivered with a factory calibration certificate and a user replaceable battery.
The set additionally includes an interface (read-write device that is required for programming the data loggers), an USB cable for connecting the logger to a PC as well as the Winlog.basic Software.
The set additionally includes an interface (read-write device that is required for programming the data loggers), an USB cable for connecting the logger to a PC as well as the Winlog.basic Software.
- Catalog No:
- 620-2837
- 620-2836
- Model:
- EBI 20-TH1
- EBI 20-TH1
- Snímač:
- NTC
- NTC
- Ochranná třída:
- IP 52
- IP 52
- Hmotnost:
- 45 g
- 45 g
- Š×H×V:
- 69×48×22 mm
- 69×48×22 mm
- Datová paměť - Počet odečtů:
- 40 000
- 40 000
- Baterie:
- 3 V Lithium
- 3 V Lithium
Frequently Bought Together