Hygiene kits for communication solutions, PELTOR™
Catalog # MMMAHY220
Dodavatel: 3M
Specifikace
- Popis:PELTOR™ hygiene kits for communication solutions
- Bal.:1 KIT
Specifikace
O tomto produktu
3M™ PELTOR™ hygiene kits communication headsets help users maintain the correct levels of protection, comfort, and hygiene.
- Extend the life of your headset
- Avoid wearing uncomfortable earmuffs
- Use the specific hygiene kit for headset maintenance
Maintain hygiene, comfort and protection of 3M™ PELTOR™ communication headsets with 3M™ PELTOR™ Hygiene Kits. Replace earmuff cushions and foam inserts every 6 months or sooner if they are damaged.