ContactUsLinkComponent
VWR®, Withdrawal System for 10 L Stainless Steel Barrels, Vertical
  99907.010
undefined
VWR®, Withdrawal System for 10 L Stainless Steel Barrels, Vertical
  99907.010
  • Popis:
    Withdrawal system for stainless steel barrels 10 L, vertical (including dip tube)
  • Materiál:
    Nerezová ocel
  • Bal.:
    1
Specially developed solvent withdrawal system for inert gas pressurizing.
  • Withdrawal system for 10 L barrels (stainless steel)
  • Perfluorelastomer o-rings
  • Includes 1 dip tube for 10 L barrels
Specially developed solvent withdrawal system for inert gas pressurizing for VWR® returnable 10 L stainless steel barrels.
 : Ideal for use with flexible hose with filling nozzle (99908.001).