96-well polyvinyl chloride (PVC) microplates
Catalog # 734-1517
Supplier: Corning
Specifications
- Doporučený pracovní objem:50 - 150 µl
- Sterilní:-
- Barva:Průhledná
- Popis:Round bottom PVC microplates, untreated
- Bal.:100
Specifications
About this item
Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
- Well volume of 250 µl (260 µl for V-bottom)
- Working well volume of 50 µl to 150 µl
Ordering information: Lids are not available.