Water stills, double distillation, Puridest
Supplier: LAUDA
Certificates
About this item
Water still (fully automatic) for double distillation, suitable for both bench and wall mounting.
- Water still (fully automatic) for double distillation, suitable for both bench and wall mounting
- Main switch with pilot lamp for the monitoring of both distillation stages at the front of the unit
- Valve made of borosilicate glass 3.3 with PTFE plunger for single distillate
- Outlet with dust guard shield made of borosilicate glass 3.3 for double distillate
- Condenser (cooling coil) 1st stage made of stainless steel
- Condenser (cooling coil) 2nd stage including the baffle made of DURAN® borosilicate glass 3.3
- Solenoid valve to control the water supply
- Economical energy consumption by distillation of the heated cooling water
- Water saving through automatic water cut-off; unnecessary water consumption will be avoided
Evaporator easily accessible for cleaning purposes with CO₂ degassing through outlet in the condenser. Electronic contamination monitoring of the still with distillation stop and visual cleaning instruction as well as electronic level control during the whole distilling process.
Double-action low water cut-off through float switch and thermostatic safety switch. Heating element and evaporator are made of stainless steel with double-walled housing made of galvanised sheet steel, powder coated.
Accessories information: Additional accessories available: prefilter, dechlorite filter, phosphate cartridge, replacement cartridge, wall mounting, hose sets.
Specifications
- Catalog No:
- 171-1315
- 171-1322
- 171-1323
- 171-1321
- 171-1332
- 171-1312
- 171-1331
- 171-1334
- 171-1313
- Výstup:
- 4 L/h
- 8 L/h
- 4 L/h
- 4 L/h
- 8 L/h
- 4 L/h
- 4 L/h
- 4 L/h
- 8 L/h
- Vodivost:
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
- Okolní teplota:
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- Výkon:
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- 400 V; 3/N/PE; 50/60 Hz
- Š×H×V:
- 650×365×390 mm
- 700×390×700 mm
- 650×365×390 mm
- 550×280×570 mm
- 700×390×700 mm
- 550×280×570 mm
- 550×280×570 mm
- 650×365×390 mm
- 700×390×700 mm
- Hmotnost:
- 28 kg
- 45 kg
- 28 kg
- 28 kg
- 45 kg
- 28 kg
- 28 kg
- 26 kg
- 48 kg
Specifications
Related Information
Frequently Bought Together