Unions for hplc
Katalogové číslo MICS49302-03
Dodavatel: MICROSOLV
Specifikace
- Popis:Union for HPLC, body only, Hole size: 0,254 mm
- Materiál:PEEK
- Bal.:1
Specifikace
O tomto produktu
The endure™ brand of carbon peek high pressure fittings are designed for extra durability. These fittings will endure higher temperatures, solvent exposure and over-tightening longer than standard peek fittings.
- Low dead volume