MacConkey Agar without CV and without NaCl, HiVeg™
Agar without CV and NaCl, 500 g
Katalogové číslo HIMEM1582-500G
Dodavatel: HIMEDIA
MacConkey Agar without CV and without NaCl, HiVeg™
Katalogové číslo HIMEM1582-500G
Dodavatel: HIMEDIA
Číslo CAS:
Přejít na:
Product Details & Documents
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based media designed to work in conjunction with or replace standard animal-based protein sources.
The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of media can meet the requirements of nearly any microbiology application.
Specifikace
Specifikace
- Bal.:500 g
- Popis:Agar without CV and NaCl