Orders LinkContactUsLinkComponent
Pouring system for 7 L Bottle
Pouring system for 7 L Bottle
Pouring system with self-closing valve, for 7 L bottle
Katalogové číslo HONC69813H-1EA
Dodavatel:   Honeywell Chemicals
Pouring system for 7 L Bottle
Katalogové číslo HONC69813H-1EA
Dodavatel:  Honeywell Chemicals
Pouring system for 7 L Bottle
Pouring system with self-closing valve, for 7 L bottle
Přejít na:
Price
2 470,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Pouring system with self-closing valve, for 7 L bottles.
Specifikace
  • Popis:
    Pouring system with self-closing valve, for 7 L bottle