Order Entry
Czech Republic
ContactUsLinkComponent
Carboxin Analytical standard, reference material, Fluka™
Carboxin Analytical standard, reference material, Fluka™
Catalog # HONCR1988-100MG
CAS Number:  
Carboxin Analytical standard, reference material, Fluka™
Catalog # HONCR1988-100MG
CAS Number:  

Specifications

  • Typ balení:
    Glass Bottle
  • Bal.:
    100 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    5234-68-4
  • Popis:
    Carboxin
  • MDL čí.:
    MFCD00055403
  • Synonym(s):
    5,6-Dihydro-2-methyl-N-phenyl-1,4-oxathiin-3-carboxamide; Carbathiin
  • Bod tání:
    90 °C
  • Vzorec:
    C₁₂H₁₃NO₂S
  • Molekulární hmotnost:
    235.30 g/mol
  • Popis:
    Carboxin
  • Fyzická forma:
    Solid

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery