Pyrimidifen Analytical standard, reference material, Fluka™
Katalogové číslo HONCR1985-10MG
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Typ balení:Glass Bottle
- Bal.:10 mg
- Standard:Analytical standard
- Čí. CAS:105779-78-0
- Popis:Pyrimidifen
- MDL čí.:MFCD01631162
- Č. UN:2811
- Synonym(s):5-Chloro-N-{2-[4-(2-ethoxyethyl)-2,3-dimethylphenoxy]ethyl}-6-ethylpyrimidin-4-amine
- Bod tání:68 °C
- Vzorec:C₂₀H₂₈ClN₃O₂
- Molekulární hmotnost:377.91 g/mol
- Popis:Pyrimidifen
- Fyzická forma:Solid
Specifikace
O tomto produktu
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery