Propamocarb Analytical standard, reference material, Fluka™
HONCR1982-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:24579-73-5
- Popis:Propamocarb
- MDL čí.:MFCD00078717
- Synonym(s):Propyl N-[3-(dimethylamino)propyl]carbamate
- Vzorec:C₉H₂₀N₂O₂
- Molekulární hmotnost:188.27 g/mol
- Popis:Propamocarb
- Bod vzplanutí:> 100 °C
- Fyzická forma:Liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery