Azinphos-ethyl Analytical standard, reference material, Fluka™
HONCR1979-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:2642-71-9
- Popis:Azinphos-ethyl
- MDL čí.:MFCD00055539
- Č. UN:2811
- Synonym(s):O,O-Diethyl S-[(4-oxo-1,2,3-benzotriazin-3(4H)-yl)methyl] phosphorodithioate
- Bod tání:49 °C
- Vzorec:C₁₂H₁₆N₃O₃PS₂
- Molekulární hmotnost:345.38 g/mol
- Popis:Azinphos-ethyl
- Fyzická forma:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery