Flumequine Analytical standard, reference material, Fluka™
HONCR1975-100MG
:
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:42835-25-6
- Popis:Flumequine
- MDL čí.:MFCD00079298
- Synonym(s):7-Fluoro-12-methyl-4-oxo-1-azatricyclo[7.3.1.05,13]trideca-2,5,7,9(13)-tetraene-3-carboxylic acid
- Bod tání:256 °C
- Vzorec:C₁₄H₁₂FNO₃
- Molekulární hmotnost:261.25 g/mol
- Popis:Flumequine
- Fyzická forma:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery