Propoxur Analytical standard, reference material, Fluka™
Katalogové číslo HONCR1972-100MG
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:114-26-1
- Popis:Propoxur
- MDL čí.:MFCD00055455
- Č. UN:2811
- Synonym(s):2-(1-Methylethoxy)phenyl N-methylcarbamate
- Bod tání:88 °C
- Vzorec:C₁₁H₁₅NO₃
- Molekulární hmotnost:209.24 g/mol
- Popis:Propoxur
- Fyzická forma:Crystalline
Specifikace
O tomto produktu
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery