Nitenpyram Analytical standard, reference material, Fluka™
Catalog # HONCR1971-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:150824-47-8
- Popis:Nitenpyram
- MDL čí.:MFCD01631161
- Synonym(s):(1E)-N-[(6-chloro-3-pyridinyl)methyl]-N-ethyl-N′-methyl-2-nitro-1,1-ethenediamine
- Bod tání:82 °C
- Vzorec:C₁₁H₁₅ClN₄O₂
- Molekulární hmotnost:270.72 g/mol
- Popis:Nitenpyram
- Fyzická forma:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery