Dicamba Analytical standard, Fluka™
Catalog # HONCR1941-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:1918-00-9
- Popis:Dicamba Analytical standard
- Podmínky uskladnění:RT
- MDL čí.:MFCD00055283
- Synonym(s):3,6-Dichloro-2-methoxybenzoic acid
- Bod tání:115 °C
- Vzorec:C₈H₆Cl₂O₃
- Molekulární hmotnost:221.04 g/mol
- Popis:Dicamba
- Fyzická forma:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery