1-Butanol Analytical standard, reference material, Fluka™
Catalog # HONCR1928-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Typ balení:Glass Bottle
- Bal.:1 g
- Standard:Analytical standard
- Čí. CAS:71-36-3
- Popis:1-Butanol
- Podmínky uskladnění:RT
- MDL čí.:MFCD00002964
- Č. UN:1120
- Synonym(s):Butyl alcohol
- Bod tání:< -90 °C
- Vzorec:C₄H₁₀O
- Molekulární hmotnost:74.12 g/mol
- Popis:1-Butanol
- Bod vzplanutí:35 °C.1,013 hPa
- Fyzická forma:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery