Climbazole Analytical standard, reference material, Fluka™
Katalogové číslo HONCR1892-100MG
Dodavatel: Honeywell Chemicals
Číslo CAS:
Specifikace
- Typ balení:Glass Bottle
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:38083-17-9
- Popis:Climbazole
- Podmínky uskladnění:RT
- MDL čí.:MFCD00055505
- Synonym(s):1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethylbutan-2-one
- Bod tání:96.6 °C
- Vzorec:C₁₅H₁₇ClN₂O₂
- Molekulární hmotnost:292.76 g/mol
- Popis:Climbazole
- Fyzická forma:Solid
Specifikace
O tomto produktu
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery